Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRINP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014618.2 | NP_055433.2 | 8 | 2032 | ||
GenBank mRNA | AF027734.1 | - | 8 | 1994 | ||
AK293501.1 | - | 2 | 550 | |||
AK293649.1 | - | 4 | 1082 | |||
BC065196.1 | - | 8 | 1985 | 5245480 | ||
BC071702.1 | - | 4 | 980 | 5244680 |