Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIFM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198696.1 | NP_001185625.1 | 7 | 962 | ||
NM_032797.5 | NP_116186.1 | 7 | 959 | |||
GenBank mRNA | AF337957.1 | - | 6 | 763 | ||
AF506757.1 | - | 7 | 959 | |||
AK027403.1 | - | 7 | 838 | |||
AK127353.1 | - | 7 | 2714 | |||
BC006121.1 | - | 6 | 767 | 3506309 | ||
BC023601.2 | - | 7 | 946 | 4548458 | ||
BX537621.1 | - | 4 | 1596 |