Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032834.3 | NP_116223.3 | 3 | 1000 | ||
GenBank mRNA | AJ312278.1 | - | 3 | 1000 | ||
AK027657.1 | - | 1 | 238 | |||
AK292488.1 | - | 3 | 766 | |||
BC033730.1 | - | 3 | 781 | |||
BC070347.1 | - | 3 | 785 | 3839674 | ||
BC090948.1 | - | 3 | 767 | 5205402 |
Gene Symbol: ALG10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013620.3 | NP_001013642.1 | 3 | 820 | ||
XM_005268665.4 | XP_005268722.1 | 3 | 601 | |||
GenBank mRNA | AK307955.1 | - | 3 | 775 | ||
AY845858.1 | - | 3 | 681 | |||
BC137413.1 | - | 3 | 736 | 9021411 | ||
BC137414.1 | - | 3 | 736 | 9021412 | ||
BC144617.1 | - | 3 | 710 |