Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRYD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032840.2 | NP_116229.1 | 2 | 216 | ||
GenBank mRNA | AK027706.1 | - | 2 | 150 | ||
AK074694.1 | - | 2 | 182 | |||
AK294092.1 | - | 2 | 200 | |||
BC136312.1 | - | 2 | 149 | 9020299 | ||
DA389433.1 | - | 2 | 216 |