Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RITA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286215.1 | NP_001273144.1 | 3 | 1519 | ||
NM_032848.2 | NP_116237.1 | 4 | 1651 | |||
XM_005253972.2 | XP_005254029.1 | 4 | 1623 | |||
GenBank mRNA | AK027733.1 | - | 4 | 1427 | ||
AK027741.1 | - | 5 | 1427 | |||
AK075358.1 | - | 4 | 1617 | |||
AK123762.1 | - | 5 | 1522 | |||
AY007158.1 | - | 4 | 1439 | |||
BC022092.2 | - | 4 | 1182 | 3029785 |