Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEDAG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032849.3 | NP_116238.2 | 4 | 1075 | ||
XM_017020801.1 | XP_016876290.1 | 3 | 1455 | |||
GenBank mRNA | AB055407.1 | - | 3 | 461 | ||
AK055635.1 | - | 4 | 1059 | |||
BC040165.1 | - | 4 | 959 | 5590156 | ||
BC101624.1 | - | 4 | 813 | 8069130 | ||
BC101626.1 | - | 4 | 813 | 8069132 |