Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032856.3 | NP_116245.2 | 3, 4 | 271 | ||
NR_130944.1 | 2, 3 | 708 | ||||
NR_130945.1 | 3, 4 | 271 | ||||
NR_130946.1 | 3, 4 | 278 | ||||
GenBank mRNA | AF161382.1 | - | 3, 4 | 202 | ||
AK027794.1 | - | 3, 4 | 208 | |||
AK055578.1 | - | 2, 3 | 708 | |||
AK055729.1 | - | 3, 4 | 208 | |||
AK090406.1 | - | 2, 3 | 1404 | |||
AK225068.1 | - | 3, 4 | 211 | |||
AK225199.1 | - | 3, 4 | 208 | |||
AK295372.1 | - | 3, 4 | 202 | |||
BC014115.2 | - | 3, 4 | 209 | |||
BC020252.2 | - | 2, 3 | 160 | |||
BC050648.1 | - | 3, 4 | 228 | |||
BC063392.1 | - | 3, 4 | 239 | 5168550 | ||
BU601517.1 | - | 281 | ||||
BX446522.2 | - | 3, 4 | 201 | |||
CN481562.1 | - | 3, 4 | 207 |