Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252335.1 | NP_001239264.1 | 3 | 1703 | ||
NM_032866.4 | NP_116255.2 | 2 | 1588 | |||
XM_005254726.3 | XP_005254783.1 | 2 | 1606 | |||
XM_005254727.4 | XP_005254784.1 | 3 | 1984 | |||
XM_011522120.1 | XP_011520422.1 | 3 | 1642 | |||
XM_011522121.1 | XP_011520423.1 | 2 | 1765 | |||
XM_017022685.1 | XP_016878174.1 | 2 | 2223 | |||
XM_017022686.1 | XP_016878175.1 | 2 | 2221 | |||
GenBank mRNA | AB051536.2 | - | 1 | 747 | ||
AK056673.1 | - | 2 | 1588 | |||
AK307410.1 | - | 2 | 1588 | |||
AY274808.1 | - | 2 | 1586 | |||
AY610514.1 | - | 3 | 1683 | |||
BC030995.1 | - | 2 | 1586 | |||
BC062770.1 | - | 1 | 978 | |||
BC093827.1 | - | 1 | 1530 | 7939672 | ||
BC112049.1 | - | 1 | 1542 | 8327517 | ||
BC118918.1 | - | 1 | 1529 | 40074275 |