Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MICALL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033386.3 | NP_203744.1 | 10 | 2275 | ||
XM_005261791.3 | XP_005261848.1 | 10 | 2088 | |||
XM_005261792.3 | XP_005261849.1 | 10 | 2082 | |||
XM_011530471.2 | XP_011528773.1 | 11 | 2259 | |||
XM_011530472.2 | XP_011528774.1 | 11 | 2253 | |||
XM_011530473.2 | XP_011528775.1 | 11 | 2259 | |||
XM_011530474.2 | XP_011528776.1 | 11 | 2670 | |||
XM_011530475.2 | XP_011528777.1 | 10 | 1804 | |||
XM_017029026.1 | XP_016884515.1 | 9 | 1821 | |||
GenBank mRNA | AB051455.1 | - | 8 | 1785 | ||
AJ496196.1 | - | 10 | 2067 | |||
AK056603.1 | - | 4 | 971 | |||
AK127819.1 | - | 1 | 99 | |||
AL833860.1 | - | 5 | 1384 | |||
AL834373.1 | - | 10 | 1932 | |||
BC001090.2 | - | 2 | 463 | |||
BC082243.1 | - | 2 | 463 | |||
CR456437.1 | - | 10 | 2006 | |||
CR749566.1 | - | 3 | 1105 |