Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM207A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316983.1 | NP_001303912.1 | 3 | 365 | ||
NM_001316984.1 | NP_001303913.1 | 3 | 320 | |||
NM_001316985.1 | NP_001303914.1 | 3 | 320 | |||
NM_001316986.1 | NP_001303915.1 | 3 | 365 | |||
NM_001316987.1 | NP_001303916.1 | 3 | 320 | |||
NM_001316988.1 | NP_001303917.1 | 3 | 303 | |||
NM_058190.3 | NP_478070.1 | 3 | 365 | |||
XM_017028481.1 | XP_016883970.1 | 3 | 513 | |||
XM_017028482.1 | XP_016883971.1 | 2 | 253 | |||
GenBank mRNA | AF391113.1 | - | 3 | 346 | ||
AF391114.1 | - | 3 | 301 | |||
BC009341.2 | - | 3 | 322 | 4128732 | ||
BG479572.1 | - | 3 | 282 | |||
BM042090.1 | - | 3 | 305 | |||
CN368049.1 | - | 3 | 353 | |||
CR998572.1 | - | 3 | 293 | |||
HY097926.1 | - | 3 | 323 |