Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033031.2 | NP_149020.2 | 8 | 3708 | ||
NM_033670.2 | NP_391990.1 | 4 | 396 | |||
XM_017029913.1 | XP_016885402.1 | 10 | 3919 | |||
XM_017029914.1 | XP_016885403.1 | 10 | 3849 | |||
XM_017029915.1 | XP_016885404.1 | 11 | 3879 | |||
XM_017029916.1 | XP_016885405.1 | 10 | 3916 | |||
XM_017029917.1 | XP_016885406.1 | 9 | 3895 | |||
XM_017029918.1 | XP_016885407.1 | 9 | 3892 | |||
GenBank mRNA | AJ314764.1 | - | 8 | 3712 | ||
AJ314765.1 | - | 4 | 400 | |||
AJ314766.1 | - | 7 | 720 | |||
AJ416458.1 | - | 8 | 3724 | |||
AK292454.1 | - | 9 | 3881 | |||
AY927611.1 | - | 1 | 573 |