Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIXDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037954.3 | NP_001033043.1 | 17 | 2014 | ||
NM_033425.4 | NP_219493.1 | 13 | 1547 | |||
XM_017018466.1 | XP_016873955.1 | 18 | 1974 | |||
XM_017018467.1 | XP_016873956.1 | 18 | 1842 | |||
XM_017018468.1 | XP_016873957.1 | 13 | 1158 | |||
XM_017018469.1 | XP_016873958.1 | 14 | 1183 | |||
GenBank mRNA | AB051522.1 | - | 13 | 1454 | ||
AK055469.1 | - | 4 | 326 | |||
AK055899.1 | - | 11 | 1070 | |||
AK289807.1 | - | 13 | 1523 | |||
AK294954.1 | - | 8 | 713 | |||
BC033034.2 | - | 13 | 1506 | 4820652 | ||
BC041626.1 | - | 10 | 741 | |||
DQ642016.1 | - | 17 | 2014 |