Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBD3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145208.2 | NP_660209.2 | 1 | 569 | ||
GenBank mRNA | AB451462.1 | - | 1 | 483 | ||
AY038022.1 | - | 1 | 506 | |||
BC112058.1 | - | 1 | 569 | 8327526 | ||
BC112064.1 | - | 1 | 569 | 8327532 |