Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UMODL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004416.2 | NP_001004416.2 | 8 | 1277 | ||
NM_001199527.1 | NP_001186456.1 | 8 | 1457 | |||
NM_001199528.2 | NP_001186457.2 | 8 | 1457 | |||
NM_173568.3 | NP_775839.3 | 8 | 1277 | |||
XM_011529797.1 | XP_011528099.1 | 6 | 885 | |||
XM_017028506.1 | XP_016883995.1 | 8 | 1272 | |||
XM_017028507.1 | XP_016883996.1 | 8 | 4201 | |||
GenBank mRNA | AB051810.1 | - | 8 | 1457 | ||
AB051811.1 | - | 8 | 1457 | |||
AB096966.1 | - | 4 | 705 | |||
AB096967.1 | - | 6 | 881 | |||
AB096968.1 | - | 6 | 870 | |||
AB096969.1 | - | 6 | 894 | |||
AB096970.1 | - | 6 | 938 | |||
AB096971.1 | - | 7 | 1050 | |||
AY771619.1 | - | 8 | 1277 | |||
AY771620.1 | - | 8 | 1277 |