Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083536.1 | NP_001077005.1 | 5 | 1226 | ||
NM_001286993.1 | NP_001273922.1 | 5 | 1208 | |||
NM_033086.2 | NP_149077.2 | 5 | 1126 | |||
XM_017015274.1 | XP_016870763.1 | 5 | 968 | |||
GenBank mRNA | AK000004.1 | - | 4 | 803 | ||
AK225843.1 | - | 5 | 878 | |||
AK292676.1 | - | 5 | 1208 | |||
AY211386.1 | - | 5 | 1062 | |||
BC032429.1 | - | 5 | 945 | 5244790 | ||
BC111054.1 | - | 5 | 1063 | 5751870 | ||
BX647829.1 | - | 5 | 1081 |