Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCHSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033449.2 | NP_258260.1 | 16 | 1609 | ||
XM_005268524.4 | XP_005268581.1 | 16 | 1636 | |||
XM_006714803.3 | XP_006714866.1 | 15 | 1513 | |||
XM_011537698.2 | XP_011536000.1 | 16 | 1642 | |||
XM_011537700.2 | XP_011536002.1 | 16 | 1589 | |||
XM_017010013.1 | XP_016865502.1 | 16 | 1642 | |||
GenBank mRNA | AK000007.1 | - | 16 | 1593 | ||
AY217346.1 | - | 16 | 1609 | |||
AY358478.1 | - | 7 | 1227 | |||
BC047016.1 | - | 15 | 1532 | 5215830 |