Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033329.2 | NP_201586.1 | 3, 4 | 810 | ||
NM_053003.3 | NP_443729.1 | 4, 5 | 1200 | |||
XM_011527441.2 | XP_011525743.1 | 4, 5 | 897 | |||
XM_011527442.2 | XP_011525744.1 | 4, 5 | 1200 | |||
GenBank mRNA | AF282256.1 | - | 4, 5 | 1180 | ||
AK095919.1 | - | 4, 5 | 1200 | |||
AK225931.1 | - | 4, 5 | 863 | |||
AY358140.1 | - | 4, 5 | 1158 | |||
BC035809.1 | - | 4, 5 | 1197 | 5755526 |