Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA5B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320373.1 | NP_001307302.1 | 9 | 1186 | ||
NM_138345.2 | NP_612354.1 | 11 | 1777 | |||
XM_011513288.2 | XP_011511590.1 | 12 | 2413 | |||
XM_011513289.2 | XP_011511591.1 | 12 | 2496 | |||
XM_011513290.2 | XP_011511592.1 | 12 | 1959 | |||
XM_011513291.2 | XP_011511593.1 | 11 | 2625 | |||
XM_011513292.2 | XP_011511594.1 | 11 | 2613 | |||
GenBank mRNA | AK293885.1 | - | 9 | 1186 | ||
AL834499.1 | - | 5 | 609 | |||
BC001200.1 | - | 6 | 956 | |||
BC136609.1 | - | 11 | 1777 | 9020599 |