Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEACAM21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098506.3 | NP_001091976.3 | 2 | 468 | ||
NM_001288773.2 | NP_001275702.2 | 3 | 718 | |||
NM_001290113.1 | NP_001277042.1 | 2 | 468 | |||
NM_033543.5 | NP_291021.4 | 2 | 468 | |||
XM_005278396.4 | XP_005278453.1 | 2 | 670 | |||
XM_005278397.3 | XP_005278454.1 | 2 | 668 | |||
XM_011527450.1 | XP_011525752.1 | 3 | 756 | |||
XM_011527452.1 | XP_011525754.1 | 3 | 718 | |||
XM_017027430.1 | XP_016882919.1 | 2 | 1042 | |||
XM_017027431.1 | XP_016882920.1 | 2 | 1042 | |||
XM_017027432.1 | XP_016882921.1 | 2 | 1040 | |||
XM_017027433.1 | XP_016882922.1 | 3 | 718 | |||
XM_017027434.1 | XP_016882923.1 | 2 | 683 | |||
XM_017027435.1 | XP_016882924.1 | 2 | 683 | |||
XM_017027436.1 | XP_016882925.1 | 6 | 1865 | |||
XM_017027437.1 | XP_016882926.1 | 2 | 2898 | |||
GenBank mRNA | AK023602.1 | - | 3 | 718 | ||
AY358084.1 | - | 2 | 412 | |||
BC012001.1 | - | 2 | 468 | 4540619 | ||
BC106727.1 | - | 2 | 398 | 40015070 | ||
BI833768.1 | - | 2 | 468 | |||
BI913326.1 | - | 3 | 710 |