Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316968.1 | NP_001303897.1 | 3 | 1063 | ||
NM_001316969.1 | NP_001303898.1 | 3 | 779 | |||
NM_033211.3 | NP_149988.1 | 3 | 823 | |||
GenBank mRNA | AF038182.1 | - | 3 | 717 | ||
AK058020.1 | - | 3 | 759 | |||
AK090558.1 | - | 3 | 995 | |||
BC009203.2 | - | 3 | 784 | 3628991 |