Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYRM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293735.1 | NP_001280664.1 | 2 | 166 | ||
NM_181705.3 | NP_859056.2 | 2 | 166 | |||
NR_121658.1 | 2 | 166 | ||||
GenBank mRNA | AK000803.1 | - | 2 | 137 | ||
BC037930.1 | - | 2 | 118 | 5284728 | ||
BC047079.1 | - | 2 | 96 | 5286889 | ||
BG714193.1 | - | 2 | 107 | |||
BI601105.1 | - | 2 | 142 | |||
BI667583.1 | - | 2 | 141 | |||
DA996312.1 | - | 2 | 167 |