Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182491.2 | NP_872297.2 | 2, 3 | 360 | ||
XM_017012774.1 | XP_016868263.1 | 2, 3 | 360 | |||
GenBank mRNA | AK027118.1 | - | 2, 3 | 302 | ||
AK290170.1 | - | 2, 3 | 319 | |||
BC016963.1 | - | 2, 3 | 295 | |||
BC029558.1 | - | 2, 3 | 303 | 5263788 |