Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HN1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144570.2 | NP_653171.1 | 4 | 408 | ||
GenBank mRNA | AK023154.1 | - | 4 | 406 | ||
AK296998.1 | - | 5 | 496 | |||
AK303927.1 | - | 4 | 313 | |||
AK304025.1 | - | 4 | 490 | |||
AK309909.1 | - | 3 | 280 | |||
AK309930.1 | - | 3 | 229 | |||
AK316514.1 | - | 4 | 464 | |||
AY323978.1 | - | 3 | 330 | |||
AY323979.1 | - | 4 | 418 | |||
AY598330.1 | - | 4 | 372 | |||
BC014438.1 | - | 4 | 383 | 4877757 | ||
BC060853.1 | - | 4 | 402 | 5296086 | ||
BM837125.1 | - | 4 | 429 |