Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHX57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198963.1 | NP_945314.1 | 23 | 3976 | ||
XM_011533154.2 | XP_011531456.1 | 23 | 3980 | |||
XM_011533155.2 | XP_011531457.1 | 23 | 3978 | |||
XM_011533156.2 | XP_011531458.1 | 23 | 3980 | |||
GenBank mRNA | AB209249.1 | - | 17 | 2328 | ||
AF070590.1 | - | 8 | 989 | |||
AF283512.1 | - | 19 | 2667 | |||
BC033636.1 | - | 4 | 346 | |||
BC039270.1 | - | 22 | 3806 | |||
BC053623.1 | - | 17 | 2277 | |||
BC065278.1 | - | 18 | 2471 | |||
BC131534.1 | - | 23 | 3948 | 40082400 |