Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139159.4 | NP_631898.3 | 15 | 1930 | ||
XM_005259673.2 | XP_005259730.1 | 16 | 2039 | |||
XM_011528405.1 | XP_011526707.1 | 17 | 2091 | |||
XM_011528406.1 | XP_011526708.1 | 18 | 2201 | |||
XM_011528408.1 | XP_011526710.1 | 15 | 1876 | |||
XM_011528410.1 | XP_011526712.1 | 16 | 2091 | |||
XM_017027453.1 | XP_016882942.1 | 17 | 2681 | |||
XM_017027454.1 | XP_016882943.1 | 18 | 2791 | |||
GenBank mRNA | AF452102.2 | - | 15 | 1950 | ||
AF542510.2 | - | 15 | 1909 | |||
AK054656.1 | - | 4 | 470 | |||
AK122645.1 | - | 9 | 980 | |||
AK122654.1 | - | 15 | 1933 | |||
AK131100.1 | - | 12 | 1499 | |||
AK131499.1 | - | 14 | 1443 | |||
AL834376.1 | - | 9 | 961 | |||
AY172660.1 | - | 12 | 1595 | |||
AY374518.1 | - | 12 | 1585 | |||
BC000970.1 | - | 3 | 313 | |||
BC037948.2 | - | 15 | 1867 | 4577856 | ||
BF203783.1 | - | 67 | ||||
CR627380.1 | - | 7 | 705 | |||
DQ417928.1 | - | 13 | 1672 |