Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139159.4 | NP_631898.3 | 22 | 2895 | ||
XM_005259673.2 | XP_005259730.1 | 23 | 3004 | |||
XM_011528405.1 | XP_011526707.1 | 24 | 3056 | |||
XM_011528406.1 | XP_011526708.1 | 25 | 3166 | |||
XM_011528408.1 | XP_011526710.1 | 22 | 2841 | |||
XM_011528410.1 | XP_011526712.1 | 23 | 3056 | |||
XM_017027453.1 | XP_016882942.1 | 24 | 3646 | |||
XM_017027454.1 | XP_016882943.1 | 25 | 3756 | |||
GenBank mRNA | AF452102.2 | - | 22 | 2915 | ||
AF542510.2 | - | 22 | 2874 | |||
AK054656.1 | - | 11 | 1435 | |||
AK075030.1 | - | 4 | 496 | |||
AK122645.1 | - | 16 | 1945 | |||
AK122654.1 | - | 22 | 2898 | |||
AK131100.1 | - | 19 | 3415 | |||
AK131499.1 | - | 21 | 2408 | |||
AY172660.1 | - | 19 | 2560 | |||
AY374518.1 | - | 19 | 2550 | |||
BC000970.1 | - | 10 | 1278 | |||
BC037948.2 | - | 22 | 2832 | 4577856 | ||
CR627380.1 | - | 14 | 1670 | |||
DQ417928.1 | - | 20 | 2637 |