Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142301.1 | NP_001135773.1 | 7 | 614 | ||
NM_153704.5 | NP_714915.3 | 6 | 683 | |||
NR_024522.1 | 6 | 683 | ||||
XM_006716686.3 | XP_006716749.1 | 5 | 521 | |||
GenBank mRNA | AK092244.1 | - | 7 | 571 | ||
AK094935.1 | - | 6 | 607 | |||
AK308071.1 | - | 7 | 573 | |||
BC032835.2 | - | 6 | 601 | 4825770 | ||
BC054338.1 | - | 6 | 610 | |||
BX648768.1 | - | 6 | 627 | |||
KU178827.1 | - | 6 | 582 | |||
KU178828.1 | - | 6 | 582 |