Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005261890.4 | XP_005261947.1 | 5 | 646 | ||
XM_006724392.3 | XP_006724455.1 | 9 | 1354 | |||
XM_006724395.3 | XP_006724458.1 | 6 | 806 | |||
XM_006724396.2 | XP_006724459.1 | 2 | 184 | |||
XM_011530621.2 | XP_011528923.1 | 2 | 171 | |||
XM_017029179.1 | XP_016884668.1 | 10 | 2035 | |||
XM_017029180.1 | XP_016884669.1 | 9 | 1493 | |||
XM_017029181.1 | XP_016884670.1 | 9 | 1931 | |||
XM_017029182.1 | XP_016884671.1 | 10 | 2035 | |||
XM_017029183.1 | XP_016884672.1 | 5 | 622 | |||
XM_017029184.1 | XP_016884673.1 | 2 | 194 | |||
XM_017029185.1 | XP_016884674.1 | 11 | 2897 | |||
XM_017029186.1 | XP_016884675.1 | 14 | 2498 |
Gene Symbol: GGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288833.1 | NP_001275762.1 | 12 | 1892 | ||
NM_013421.2 | NP_038265.2 | 13 | 1820 | |||
NM_013430.2 | NP_038347.2 | 12 | 1580 | |||
GenBank mRNA | AL832738.1 | - | 1796 | |||
BC025927.1 | - | 10 | 1460 | 4548861 | ||
BC065921.1 | - | 2 | 103 | |||
BC069326.1 | - | 1 | 61 | |||
BC069473.1 | - | 1 | 61 | |||
BC069504.1 | - | 1 | 61 | |||
BC128234.1 | - | 1 | 89 | |||
BC128238.1 | - | 1 | 90 | |||
BC128239.1 | - | 1 | 90 | |||
CR456494.1 | - | 9 | 1170 | |||
DQ000492.2 | - | 3 | 275 | |||
J04131.1 | - | 13 | 1762 | |||
J05235.1 | - | 11 | 1859 | |||
L20490.1 | - | 2 | 100 | |||
L20493.1 | - | 1 | 249 | |||
M24087.1 | - | 8 | 1095 | |||
M24903.1 | - | 12 | 1580 | |||
X60069.1 | - | 10 | 1451 |
Gene Symbol: GGT3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003267.1 | 9 | 1287 | |||
GenBank mRNA | BC143607.1 | - | 6 | 801 |
Gene Symbol: GGTLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178311.2 | NP_842563.1 | 2 | 266 | ||
NM_178312.2 | NP_842564.1 | 2 | 194 | |||
XM_005260863.4 | XP_005260920.1 | 2 | 621 | |||
XM_005260864.4 | XP_005260921.1 | 3 | 774 | |||
XM_005260865.4 | XP_005260922.1 | 3 | 774 | |||
XM_011529391.2 | XP_011527693.1 | 2 | 188 | |||
XM_011529392.2 | XP_011527694.1 | 3 | 713 | |||
XM_011529393.2 | XP_011527695.1 | 2 | 560 | |||
XM_017028126.1 | XP_016883615.1 | 2 | 183 | |||
GenBank mRNA | BC040904.1 | - | 2 | 266 | 5744021 | |
L20491.1 | - | 2 | 194 | |||
L20492.1 | - | 1 | 244 |
Gene Symbol: GGTLC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282879.1 | NP_001269808.1 | 2 | 123 | ||
NM_199127.2 | NP_954578.2 | 1 | 95 | |||
XM_005261815.3 | XP_005261872.2 | 2 | 152 | |||
XM_005261817.2 | XP_005261874.1 | 2 | 127 | |||
XM_005261818.2 | XP_005261875.1 | 2 | 152 | |||
XM_011530497.1 | XP_011528799.1 | 2 | 235 | |||
GenBank mRNA | BC069534.1 | - | 1 | 61 | 7262125 | |
BX282400.1 | - | 2 | 125 | |||
EL736203.1 | - | 1 | 61 | |||
HY050283.1 | - | 2 | 114 | |||
X98922.1 | - | 1 | 96 |
Gene Symbol: LOC102724197 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724921.3 | XP_006724984.1 | 9 | 1797 | ||
XM_006724924.3 | XP_006724987.1 | 8 | 1639 | |||
XM_006724928.3 | XP_006724991.1 | 4 | 835 | |||
XM_006724929.2 | XP_006724992.1 | 2 | 184 | |||
XM_011546256.2 | XP_011544558.1 | 10 | 1931 | |||
XM_017030133.1 | XP_016885622.1 | 10 | 1362 | |||
XM_017030134.1 | XP_016885623.1 | 10 | 1365 | |||
XM_017030135.1 | XP_016885624.1 | 10 | 1790 | |||
XM_017030136.1 | XP_016885625.1 | 11 | 1832 | |||
XM_017030137.1 | XP_016885626.1 | 11 | 1829 | |||
XM_017030138.1 | XP_016885627.1 | 11 | 1352 | |||
XM_017030139.1 | XP_016885628.1 | 11 | 1355 | |||
XM_017030140.1 | XP_016885629.1 | 12 | 1822 | |||
XM_017030141.1 | XP_016885630.1 | 12 | 1819 | |||
XM_017030142.1 | XP_016885631.1 | 10 | 1663 | |||
XM_017030143.1 | XP_016885632.1 | 9 | 1798 | |||
XM_017030144.1 | XP_016885633.1 | 5 | 726 | |||
XM_017030145.1 | XP_016885634.1 | 6 | 1359 |
Gene Symbol: LOC102724823 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030162.1 | XP_016885651.1 | 10 | 1831 |