Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282669.1 | NP_001269598.1 | 7 | 1229 | ||
NM_001320132.1 | NP_001307061.1 | 7 | 1326 | |||
NM_001320136.1 | NP_001307065.1 | 7 | 1207 | |||
NM_004402.3 | NP_004393.1 | 7 | 1157 | |||
NR_104222.1 | 8 | 1294 | ||||
NR_135150.1 | 8 | 1281 | ||||
NR_135151.1 | 8 | 1244 | ||||
NR_135152.1 | 7 | 1179 | ||||
XM_017000498.1 | XP_016855987.1 | 6 | 1092 | |||
XM_017000499.1 | XP_016855988.1 | 6 | 1108 | |||
XM_017000500.1 | XP_016855989.1 | 6 | 863 | |||
GenBank mRNA | AB013918.1 | - | 7 | 820 | ||
AB028911.1 | - | 8 | 1074 | |||
AB028912.1 | - | 7 | 1120 | |||
AB028913.1 | - | 8 | 1038 | |||
AF039210.1 | - | 7 | 820 | |||
AF064019.1 | - | 7 | 951 | |||
AF409060.1 | - | 9 | 831 | |||
AF409061.1 | - | 7 | 846 | |||
AF409062.1 | - | 7 | 874 | |||
AK290877.1 | - | 7 | 948 | |||
AK303065.1 | - | 7 | 1036 | |||
AY438267.1 | - | 7 | 749 | |||
AY438268.1 | - | 4 | 372 | |||
BC032827.1 | - | 8 | 1106 | |||
BC048797.1 | - | 7 | 1143 | 5204996 |