Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKDCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138370.2 | NP_612379.2 | 2 | 900 | ||
GenBank mRNA | BC094697.1 | - | 2 | 308 | 30529646 | |
BC110513.1 | - | 2 | 337 | 40031755 | ||
BC110514.1 | - | 2 | 337 | 40031756 | ||
CF131963.1 | - | 2 | 396 | |||
DB231841.1 | - | 3 | 366 |