Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303508.1 | NP_001290437.1 | 3 | 974 | ||
GenBank mRNA | AB219123.1 | - | 2 | 381 | ||
AK025181.1 | - | 2 | 1263 | |||
BC112212.1 | - | 2 | 372 | 8327680 | ||
BC113633.1 | - | 2 | 360 | 8322685 | ||
CR749372.1 | - | 3 | 975 |