Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS19BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194326.3 | NP_919307.1 | 2, 3 | 295 | ||
GenBank mRNA | AK025423.1 | - | 2, 3 | 188 | ||
BC037573.1 | - | 2, 3 | 305 | 5399759 | ||
BC047711.1 | - | 2, 3 | 187 | 6064917 | ||
CR456443.1 | - | 2, 3 | 225 |