Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LONRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152271.3 | NP_689484.3 | 7 | 1588 | ||
XM_005273685.3 | XP_005273742.1 | 7 | 1557 | |||
XM_011544694.2 | XP_011542996.1 | 7 | 1340 | |||
GenBank mRNA | AA284268.1 | - | 4 | 555 | ||
AK074329.1 | - | 2 | 140 | |||
AK297266.1 | - | 6 | 725 | |||
AK300538.1 | - | 3 | 334 | |||
BC106042.1 | - | 7 | 854 | |||
BC117381.1 | - | 4 | 519 | 40125932 | ||
BC117385.1 | - | 4 | 519 | 40125936 | ||
BQ718281.1 | - | 185 |