Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005268208.1 | XP_005268265.1 | 18 | 2394 | ||
XM_005268209.3 | XP_005268266.1 | 18 | 2198 | |||
XM_017021765.1 | XP_016877254.1 | 18 | 2161 | |||
GenBank mRNA | AB082526.1 | - | 18 | 2330 | ||
AL832633.2 | - | 12 | 2018 | |||
AY048580.1 | - | 18 | 2224 | |||
AY080896.1 | - | 18 | 2236 | |||
BC001714.1 | - | 3 | 255 | |||
BC009336.2 | - | 3 | 266 | |||
BC093881.1 | - | 18 | 2305 | 7939726 | ||
BC112101.1 | - | 18 | 2305 | 8327569 |