Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317899.1 | NP_001304828.1 | 9 | 1150 | ||
NM_152272.4 | NP_689485.1 | 10 | 1864 | |||
XM_006716415.2 | XP_006716478.1 | 8 | 1259 | |||
XM_017013961.1 | XP_016869450.1 | 9 | 1320 | |||
XM_017013962.1 | XP_016869451.1 | 11 | 2001 | |||
XM_017013963.1 | XP_016869452.1 | 9 | 1127 | |||
XM_017013964.1 | XP_016869453.1 | 10 | 3013 | |||
XM_017013965.1 | XP_016869454.1 | 8 | 1066 | |||
GenBank mRNA | AK021776.1 | - | 9 | 1293 | ||
AK092500.1 | - | 9 | 1100 | |||
AK097202.1 | - | 10 | 1861 | |||
AK315664.1 | - | 9 | 1301 | |||
AL832313.1 | - | 6 | 1245 | |||
AL833843.1 | - | 9 | 1269 | |||
BC004344.1 | - | 9 | 1092 | |||
BC019110.1 | - | 9 | 1266 | 5093453 | ||
BC042050.1 | - | 10 | 1845 | 5551762 |