Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153603.3 | NP_705831.1 | 6 | 949 | ||
XM_017023870.1 | XP_016879359.1 | 6 | 992 | |||
GenBank mRNA | AK093989.1 | - | 3 | 305 | ||
BC000549.2 | - | 6 | 904 | 3163316 | ||
BC037563.1 | - | 6 | 938 | 5393265 | ||
KU178837.1 | - | 6 | 752 |