Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152277.2 | NP_689490.2 | 3 | 776 | ||
XM_017010022.1 | XP_016865511.1 | 3 | 786 | |||
XM_017010023.1 | XP_016865512.1 | 3 | 710 | |||
GenBank mRNA | AF251700.1 | - | 1 | 298 | ||
AK022894.1 | - | 1 | 306 | |||
AK313274.1 | - | 3 | 742 | |||
BC019910.1 | - | 3 | 624 | 4402070 | ||
BU849695.1 | - | 3 | 791 |