Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310338.1 | NP_001297267.1 | 2 | 354 | ||
NM_001310339.1 | NP_001297268.1 | 2 | 354 | |||
NM_052865.3 | NP_443097.1 | 2 | 354 | |||
XM_005260867.2 | XP_005260924.1 | 2 | 576 | |||
XM_005260870.3 | XP_005260927.1 | 2 | 302 | |||
XM_006723663.3 | XP_006723726.1 | 2 | 302 | |||
XM_011529394.1 | XP_011527696.1 | 2 | 191 | |||
XM_011529395.1 | XP_011527697.1 | 2 | 354 | |||
XM_017028127.1 | XP_016883616.1 | 2 | 576 | |||
XM_017028128.1 | XP_016883617.1 | 2 | 450 | |||
GenBank mRNA | AF306684.1 | - | 2 | 162 | ||
AF306685.1 | - | 2 | 162 | |||
AF306686.1 | - | 2 | 200 | |||
AK027503.1 | - | 2 | 158 | |||
AK315532.1 | - | 2 | 190 | |||
BC016869.1 | - | 2 | 126 | 3850853 | ||
BX382492.2 | - | 1 | 119 | |||
CV807140.1 | - | 2 | 167 | |||
DB014120.1 | - | 2 | 354 |