Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARRDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317968.1 | NP_001304897.1 | 5, 6 | 699 | ||
NM_152285.3 | NP_689498.1 | 5, 6 | 699 | |||
XM_005266119.1 | XP_005266176.1 | 5, 6 | 749 | |||
XM_006717320.3 | XP_006717383.1 | 4, 5 | 1121 | |||
XM_006717321.3 | XP_006717384.1 | 4, 5 | 1121 | |||
XM_006717322.3 | XP_006717385.1 | 4, 5 | 1070 | |||
XM_017015288.1 | XP_016870777.1 | 4, 5 | 1070 | |||
GenBank mRNA | AB209523.1 | - | 1, 2 | 1412 | ||
AJ420420.1 | - | 5, 6 | 662 | |||
AK001822.1 | - | 3, 4 | 1221 | |||
AK308376.1 | - | 5, 6 | 636 | |||
AL542109.3 | - | 5, 6 | 694 | |||
BC032346.1 | - | 5, 6 | 685 | 5211669 | ||
BI906381.1 | - | 5, 6 | 700 |