Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHKBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138392.3 | NP_612401.2 | 11 | 1079 | ||
XM_006723474.1 | XP_006723537.1 | 11 | 1079 | |||
XM_006723475.1 | XP_006723538.1 | 11 | 1079 | |||
XM_011527493.1 | XP_011525795.1 | 10 | 975 | |||
XM_017027473.1 | XP_016882962.1 | 11 | 1079 | |||
XM_017027474.1 | XP_016882963.1 | 10 | 975 | |||
GenBank mRNA | AF258553.1 | - | 11 | 1078 | ||
AK075057.1 | - | 11 | 1001 | |||
AK123695.1 | - | 9 | 1057 | |||
AK297006.1 | - | 5 | 705 | |||
AK300808.1 | - | 9 | 958 | |||
AK300819.1 | - | 11 | 1029 | |||
AK312748.1 | - | 11 | 1043 | |||
AY007117.1 | - | 2 | 114 | |||
BC007653.1 | - | 3 | 365 | |||
BC022855.1 | - | 11 | 1050 | 4102901 |