Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113525.1 | NP_001106997.1 | 6 | 1257 | ||
NM_152287.3 | NP_689500.2 | 6 | 1247 | |||
NR_110122.1 | 6 | 1247 | ||||
NR_110126.1 | 6 | 1215 | ||||
NR_110128.1 | 6 | 1138 | ||||
NR_110129.1 | 6 | 1227 | ||||
XM_005256324.3 | XP_005256381.1 | 6 | 1206 | |||
XM_005256328.3 | XP_005256385.1 | 4 | 887 | |||
XM_017023890.1 | XP_016879379.1 | 5 | 1182 | |||
GenBank mRNA | AK026482.1 | - | 6 | 1197 | ||
AK056405.1 | - | 6 | 1198 | |||
AK289801.1 | - | 6 | 1106 | |||
AK295770.1 | - | 4 | 919 | |||
AK315793.1 | - | 3 | 614 | |||
BC032781.1 | - | 5 | 916 | |||
BC105959.1 | - | 6 | 1040 | 40036181 | ||
BC107603.1 | - | 6 | 1072 | 40036180 | ||
BC110348.1 | - | 6 | 972 | 5196957 |