Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2Q2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145335.1 | NP_001138807.1 | 3 | 561 | ||
NM_173469.3 | NP_775740.1 | 3 | 680 | |||
XM_005254787.1 | XP_005254844.1 | 3 | 452 | |||
XM_005254788.1 | XP_005254845.1 | 3 | 452 | |||
XM_006720770.1 | XP_006720833.1 | 3 | 452 | |||
XM_011522228.1 | XP_011520530.1 | 3 | 452 | |||
XM_017022726.1 | XP_016878215.1 | 3 | 556 | |||
GenBank mRNA | AK294199.1 | - | 3 | 221 | ||
AK296234.1 | - | 3 | 561 | |||
AL832429.1 | - | 3 | 505 | |||
BC017708.1 | - | 3 | 507 | 3907760 |