Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001552.4 | NP_001001552.3 | 4 | 366 | ||
NM_001199050.1 | NP_001185979.1 | 6 | 508 | |||
NM_001199051.1 | NP_001185980.1 | 5 | 385 | |||
NM_001199052.1 | NP_001185981.1 | 3 | 243 | |||
NR_037583.1 | 4 | 320 | ||||
XM_011510160.2 | XP_011508462.1 | 7 | 596 | |||
XM_011510161.2 | XP_011508463.1 | 7 | 584 | |||
XM_011510162.2 | XP_011508464.1 | 7 | 576 | |||
XM_011510163.2 | XP_011508465.1 | 6 | 476 | |||
XM_011510164.2 | XP_011508466.1 | 8 | 663 | |||
XM_011510165.2 | XP_011508467.1 | 8 | 635 | |||
GenBank mRNA | AB084764.1 | - | 3 | 210 | ||
AB084765.1 | - | 4 | 287 | |||
AB085610.1 | - | 4 | 333 | |||
AB096677.1 | - | 6 | 475 | |||
AB096678.1 | - | 4 | 287 | |||
AB096679.1 | - | 5 | 352 | |||
AB096680.1 | - | 5 | 410 | |||
AB096681.1 | - | 4 | 333 | |||
AB096682.1 | - | 3 | 210 | |||
AK026628.1 | - | 4 | 333 | |||
AK302392.1 | - | 6 | 510 | |||
BC036636.2 | - | 6 | 478 | 5296437 | ||
DC399934.1 | - | 6 | 510 |