Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011699.1 | NP_001011699.1 | 11 | 1150 | ||
NM_033417.1 | NP_219485.1 | 11 | 1153 | |||
XM_005260154.4 | XP_005260211.1 | 10 | 986 | |||
GenBank mRNA | AK295994.1 | - | 11 | 1225 | ||
BC004398.1 | - | 11 | 1040 | |||
BC010176.2 | - | 5 | 589 | 3342874 |