Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134665.2 | NP_001128137.1 | 5, 6 | 671 | ||
NM_001134666.2 | NP_001128138.1 | 5, 6 | 660 | |||
NM_152292.4 | NP_689505.1 | 5, 6 | 816 | |||
XM_005263352.3 | XP_005263409.1 | 5, 6 | 650 | |||
XM_006714417.2 | XP_006714480.1 | 5, 6 | 692 | |||
GenBank mRNA | AF106046.1 | - | 3, 4 | 186 | ||
AK000332.1 | - | 5, 6 | 622 | |||
AK313548.1 | - | 5, 6 | 646 | |||
BC028373.2 | - | 5, 6 | 816 | 4838346 | ||
EU668342.1 | - | 5, 6 | 816 |