Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATPIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016311.4 | NP_057395.1 | 3 | 479 | ||
NM_178190.2 | NP_835497.1 | 3 | 555 | |||
NM_178191.2 | NP_835498.1 | 2 | 1863 | |||
GenBank mRNA | AF114836.1 | - | 565 | |||
AK000934.1 | - | 2 | 1800 | |||
AL050386.1 | - | 2 | 1793 | |||
AL583540.3 | - | 3 | 478 | |||
BC004955.1 | - | 3 | 362 | 3625379 | ||
BC009677.1 | - | 3 | 437 | 3877506 | ||
BG036978.1 | - | 3 | 527 | |||
BG337531.1 | - | 3 | 435 | |||
BP271519.1 | - | 3 | 475 |