Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORMDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128150.1 | NP_001121622.1 | 2 | 153 | ||
NM_016467.4 | NP_057551.1 | 3 | 263 | |||
XM_005246970.2 | XP_005247027.1 | 2 | 182 | |||
XM_011512199.2 | XP_011510501.1 | 2 | 145 | |||
XM_011512200.2 | XP_011510502.1 | 1 | 1760 | |||
XM_017005347.1 | XP_016860836.1 | 3 | 302 | |||
XM_017005348.1 | XP_016860837.1 | 2 | 134 | |||
GenBank mRNA | AB064959.1 | - | 1 | 722 | ||
AB064960.1 | - | 2 | 161 | |||
AF151036.1 | - | 2 | 101 | |||
AF395704.1 | - | 3 | 218 | |||
AK074756.1 | - | 2 | 152 | |||
AK075160.1 | - | 1 | 154 | |||
AK313533.1 | - | 3 | 228 | |||
BC005200.1 | - | 2 | 120 | 3929844 | ||
CR936635.1 | - | 1 | 830 | |||
DB122266.1 | - | 2 | 153 |