Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308103.1 | NP_001295032.1 | 3 | 392 | ||
NM_022073.3 | NP_071356.1 | 3 | 926 | |||
GenBank mRNA | AJ310545.1 | - | 3 | 926 | ||
AK025273.1 | - | 3 | 926 | |||
AK123350.1 | - | 4 | 755 | |||
AK225473.1 | - | 3 | 926 | |||
BC010992.2 | - | 3 | 882 | 3948676 | ||
BC064924.1 | - | 3 | 886 | 6141425 | ||
BC102030.1 | - | 3 | 392 | 40032049 | ||
BC105938.1 | - | 3 | 392 | 40032045 | ||
BC105939.1 | - | 3 | 674 | 40032051 | ||
BC111057.1 | - | 3 | 909 | 5747720 | ||
BX346941.2 | - | 2 | 249 | |||
DQ975379.1 | - | 3 | 326 |