Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145427.1 | NP_001138899.1 | 1 | 1844 | ||
NM_052870.2 | NP_443102.2 | 1 | 1840 | |||
GenBank mRNA | AF395536.1 | - | 1 | 1650 | ||
AK304195.1 | - | 1 | 1844 | |||
BC117218.1 | - | 1 | 1738 | 40125769 | ||
BC117220.1 | - | 1 | 1738 | 40125771 |