Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001653.2 | NP_001001653.1 | 3 | 299 | ||
NM_052877.4 | NP_443109.2 | 3 | 256 | |||
NM_201542.4 | NP_963836.2 | 3 | 256 | |||
GenBank mRNA | AF521562.1 | - | 2 | 208 | ||
BC010019.2 | - | 3 | 271 | 2905623 | ||
BC010543.1 | - | 2 | 197 | |||
BG620658.1 | - | 3 | 257 | |||
BI488866.1 | - | 2 | 212 |